missing translation for 'onlineSavingsMsg'
Learn More

BOLA Filtering Tiles

Product Code. 10185401
Change view
Click to view available options
Thickness (Metric):
1 mm
2 mm
Sample Volume:
5 μL
10 μL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thickness (Metric) Sample Volume unitSize
10185401 1 mm 10 μL Each
10125361 1 mm 5 μL Each
10398891 2 mm 10 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10185401 Supplier BOLA Supplier No. N161610
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Filtering Tiles Porous PTFE

Made of microporous PTFE, standard tiles with dimensions of 320 x 320 mm for cutting or stamping.

Specifications

Autoclavable Autoclavable
Sample Volume 10 μL
Sterility Non-sterile
Thickness (Metric) 1 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.