Learn More
CELLPATH FILOSLIDE 25 BOX
Click to view available options
Color:
Black
Blue
Orange
Red
White
Unit Size:
Each
Description
- Key Benefits:
- Stackable
- Snap closure to seal out dust
- Splash-proof
- Double hinged lid prevents loss or breakage
- Index card included for easy specimen identification
- Can store up to 25 micrscope slides
Specifications
Specifications
| Type | Slide Box |
| Slide Capacity | 25 |
| Color | Orange |