Learn More
CELLPATH FILOSLIDE 25 BOX
Product Code. 30226300
Click to view available options
Color:
Black
Blue
Orange
Red
White
Unit Size:
Each
Product Code. 30226300
Brand: CELLPATH WEQ020700A
Only null left
Add to basket
Add to basket
This sturdy FiloSlide Box offers safe and cost-effective storage for microscope slides.
  • Key Benefits:
  • Stackable
  • Snap closure to seal out dust
  • Splash-proof
  • Double hinged lid prevents loss or breakage
  • Index card included for easy specimen identification
  • Can store up to 25 micrscope slides
Specifications
Type Slide Box
Slide Capacity 25
Color Orange