Learn More
FIIN-3, MedChemExpress
Product Code. 16483547
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16483547 50 mg 50mg
16480008 5 mg 5mg
16467528 10 mg 10mg
16407486 100 mg 100mg
16401739 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16483547 Supplier Medchem Express Supplier No. HY18603
MedChemExpress FIIN-3 is an irreversible inhibitor of FGFR with an IC50 of 13.1, 21, 31.4, and 35.3 nM for FGFR1, FGFR2, FGFR3 and FGFR4, respectively.
FIIN-3 is an irreversible inhibitor of FGFR with an IC50 of 13.1, 21, 31.4, and 35.3 nM for FGFR1, FGFR2, FGFR3 and FGFR4, respectively.
Specifications
Chemical Name or Material FIIN-3
CAS 1637735-84-2
Color White
Purity Grade Notes Research
Molecular Formula C34H36Cl2N8O4
Quantity 50 mg
Solubility Information DMSO : 10 mg/mL (14.46 mM; Need ultrasonic and warming)
SMILES C=CC(NC1=CC=C(CN(C(NC2=C(Cl)C(OC)=CC(OC)=C2Cl)=O)C3=NC=NC(NC4=CC=C(N5CCN(C)CC5)C=C4)=C3)C=C1)=O
Molecular Weight (g/mol) 691.61
Formula Weight 691.61
Percent Purity 96.47%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease