missing translation for 'onlineSavingsMsg'
Learn More

FIIN-2, MedChemExpress

Product Code. p-200097005
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16473547

Brand: Medchem Express HY18602

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress FIIN-2 is an irreversible inhibitor of FGFR with an IC50 of 3.1, 4.3, 27, and 45 nM for FGFR1, FGFR2, FGFR3 and FGFR4, respectively.

FIIN-2 is an irreversible inhibitor of FGFR with an IC50 of 3.1, 4.3, 27, and 45 nM for FGFR1, FGFR2, FGFR3 and FGFR4, respectively.

Specifications

Chemical Name or Material FIIN-2
CAS 1633044-56-0
Color White
Purity Grade Notes Research
Molecular Formula C35H38N8O4
Quantity 50 mg
Solubility Information DMSO : ≥ 30 mg/mL (47.26 mM)
SMILES C=CC(NC1=CC=C(CN(C2=NC(NC3=CC=C(N4CCN(C)CC4)C=C3)=NC=C2CN5C6=CC(OC)=CC(OC)=C6)C5=O)C=C1)=O
Molecular Weight (g/mol) 634.73
Formula Weight 634.73
Percent Purity 99.11%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.