Learn More
Thermo Scientific™ Fiberlite F23-48 x 1.5 for LYNX 4000 and 6000 Superspeed Centrifuges
Description
Combine high g-force and high capacity for 1.5mL microtubes with the Thermo Scientific™ Fiberlite™ F23-48 x 1.5mL Fixed-Angle Rotor.
Features:
- Lightweight carbon fiber and innovative construction allow unique rotor designs not possible with metal
- Corrosion and fatigue-resistance secures the rotor's structural integrity for unequalled durability
- Rotor is backed by a 15-year warranty
- Secure, push-button rotor exchange in less than three seconds
- Improved safety and convenience
- No tools required
- Quickly change rotors and applications
Thermo Scientific™ Auto-ID™ Instant Rotor Identification:
- Immediate identification of a rotor
- Rotor specifications automatically loaded into centrifuge parameters
- Shortens run set-up time, improves safety
Speed Handle:
- Accelerates and simplifies rotor lid tightening, ensuring lid is properly attached
- Easier and safer lifting and carrying of rotors, further enhanced with the lightweight design
Specifications
Specifications
| Description | Fiberlite F23-48x1.5 Fixed Angle Rotor |
| Material | Carbon Fiber Composite |
| Type | Fixed Angle Rotor |
| Capacity | 48 x 1.5 mL |
| Max. RCF | 57,368 x g |
| Max. Speed | 23,000 rpm |
| Radius (Metric) | 97 mm |
| Radius (Metric) Min. | 64 mm |
| K Factor | 137 |
| Max. Tube Length | 40 mm |
| Show More |