Learn More
Thermo Scientific™ Fiberlite™ F14-6 x 250LE Rotor Adapters
Product Code. 11808372 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity:
15 mL Conical Tube (Set of 2)
5 mL Round-bottom Tube
50 mL Conical Tube (Set of 2)
50 mL Round-bottom Tube (Set of 2)
7 mL Round-bottom Tube
Unit Size:
Each
Pack of 2
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity unitSize
11808372 15 mL Conical Tube (Set of 2) Pack of 2
11874511 5 mL Round-bottom Tube Each
12796947 7 mL Round-bottom Tube Each
11844511 50 mL Round-bottom Tube (Set of 2) Pack of 2
11898362 50 mL Conical Tube (Set of 2) Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11808372 Supplier Thermo Scientific™ Supplier No. 75101073
Only null left
Add to basket
Add to basket
Combine this rotor Adapter with the Fiberlite F14-6 x 250LE fixed-angle rotor.
Combine Thermo Scientific™ Fiberlite™ F14-6 x 250LE Rotor Adapters with the Fiberlite F14-6 x 250LE fixed-angle rotor.
Note:
  • Adapters 00369 and 00456 must be used together for 7mL round-bottom tubes
Ordering Information:
Fiberlite F14-6x250LE fixed-angle rotor body (75003662) is also available. Contact your sales representative for more information.
TRUSTED_SUSTAINABILITY
Specifications
Type Rotor Adapter
Capacity 15 mL Conical Tube (Set of 2)