Learn More
SCHOTT AG Lighting and Imaging™ Fiber Optics Lightguide Adapter
GREENER_CHOICE
Product Code. 30052752 Shop All Schott Products
Click to view available options
For Use With (Equipment):
KL 1500 HAL Cold Light Source
Unit Size:
Each
Product Code. 30052752
Brand: SCHOTT AG Lighting and Imaging™ 158320
Only null left
Add to basket
Add to basket
This Fiber Optics Lightguide Adapter can be used to mount fiber optic lightguides from Volpi/Photonic on the SCHOTT Coldlightsource (KL1500HAL).
  • Lightguides from Volpi or Photonic have a different endtip diameter than the SCHOTT Fiber Optic Lightguide
  • With this adapter, Lightguides from Volpi or Photonic can be used with a SCHOTT Cold Light Source
  • For use only with SCHOTT Cold Light Source (KL1500HAL)
Specifications
Type Mount Adapter
For Use With (Equipment) KL 1500 HAL Cold Light Source