missing translation for 'onlineSavingsMsg'
Learn More

FGFBP2 Rabbit anti-Human, Polyclonal, Proteintech

Product Code. 16881374 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16881374 20 μL 20µL
16871374 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16881374 Supplier Proteintech Supplier No. 132541AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This gene encodes a member of the fibroblast growth factor binding protein family. The encoded protein is a serum protein that is selectively secreted by cytotoxic lymphocytes and may be involved in cytotoxic lymphocyte-mediated immunity. An increase in the amount of gene product may be associated with atopic asthma and mild extrinsic asthma.

Specifications

Antigen FGFBP2
Applications Immunocytochemistry, Western Blot, Immunofluorescence
Classification Polyclonal
Concentration 0.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene FGFBP2
Gene Accession No. Q9BYJ0
Gene Alias FGF binding protein 2, FGF BP2, FGFBP 2, FGFBP2, HBp17 related protein, HBp17 RP, HBP17RP, KSP37
Gene Symbols FGFBP2
Host Species Rabbit
Immunogen FGFBP2 Fusion Protein Ag4036
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83888
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.