Learn More
FGF18 Antibody (CL7628) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-44231
Product Code. 30303057
  • 638.00 EUR / 100µL
Description
FGF18 Monoclonal antibody specifically detects FGF18 in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin),Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FGF18
Monoclonal
LYOPH
Western Blot 1 μg/mL, Immunohistochemistry-Paraffin 1:500 - 1:1000
FGF-18ZFGF5, fibroblast growth factor 18, zFGF5
This antibody was generated using a synthetic peptide of O76093, with the exact immunogen sequence remaining proprietary.
100 μL
Angiogenesis, Biologically Active Proteins, Cell Cycle and Replication
8817
Human
Purified
Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry
CL7628
Unconjugated
Lyophilized in 5% Trehalose.
Mouse
Protein A purified
RUO
Primary
Centrifuge the vial of lyophilized antibody at 12,000 x g for 20 seconds. Reconstitute by adding sterile, distilled water to achieve a final antibody concentration of 1mg/mL.
Store at 4⪚C short term. Aliquot and store at -20⪚C long term. Avoid freeze-thaw cycles.
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions