Learn More
Invitrogen™ FGF13 Polyclonal Antibody

Description
Immunogen sequence: MAAAIASSLI RQKRQARERE KSNACKCVSS PSKGKTSCDK NKLNVFSRVK LFGSKKRRRR RPEPQLKGIV TKLYSRQGYH LQLQADGTID GTKDEDSTYT LFNLIPVGLR VVAIQGVQTK LYLAMNSEGY LYTSELFTPE CKFKESVFEN YYVTYSSMIY RQQQSGRGWY LGLNKEGEIM KGNHVKKNKP AAHFLPKPLK VAMYKEPSLH DLTEFSRSGS GTPTKSRSVS GVLNGGKSMS HNEST; Positive Samples: Mouse brain, Mouse heart, Mouse spinal cord, Rat brain, Rat heart; Cellular Location: Cell projection, Cytoplasm, Cytoplasm, Nucleus, dendrite, filopodium, growth cone, nucleolus.
Specifications
Specifications
| Antigen | FGF13 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.42 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | FGF13 |
| Gene Accession No. | P70377, Q92913, Q9ERW3 |
| Gene Alias | AFGF; ECGF; ECGFA; ECGFB; FGF; FGF 1; FGF alpha; FGF13; FGF-13; FGF13A; FGF2; FGFA; Fhf2; FHF-2; Fibroblast growth factor; fibroblast growth factor 13; Fibroblast growth factor homologous factor 2; GLIO703; HBGF 1; HBGF1 |
| Gene Symbols | FGF13 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.