Learn More
FFP3 Disposable Respirator Face Mask
Shop All Medicom Products
Click to view available options
:
Without Valve
Description
- Tested and certified FFP3 disposable respirator
- Without exhalation valve
- Head bands enable a more secure, tighter and comfortable fit
- Head bands make wearing glasses more comfortable and enable good compatibility with various eye protection products
Specifications
Specifications
| Certifications/Compliance | CE Approved, EN 149:2001+A1:2009 FFP3 NR |
| Design Type | Flat-fold |
| Disposable | Single-use |
| Filter Type | FFP3 NR |
| Material | Face Piece: Polypropylene; Head Band: Synthetic Elastic; Nosepiece: Metal and Polypropylene |
| Product Type | Particulate Respirator with Double Head Bands |
| Size | One Size |
| With Exhalation Valve | No |
| Packaging Type | Individually Packed |
| Sterility | Non-sterile |