Learn More
Chemsplash FFP3 Cup Mask with Valve
Description
Chemsplash 3492 Valved Cup Dust Mask
- High Filtration Capacity
- Red Color Exhalation Valve
- Exhalation Valve Reduces Hot Air Buildup
- Full Inner Face Seal with Excellent Fit
- Plastic Welded Woven Elasticated Adjustable Head Straps
- Protects Against Harmful and Toxic Solid and Liquid Particles
- No Vision Impairment and Problem Free Speech
- Latex and Graphene Free
Description: The Chemsplash 3492 is a Valved Cup Dust Mask, colour coded red for ease of use. The fabric material of the mask has high filtering efficiency, and it features plastic welded woven elasticated adjustable head straps for a secure fit. It also provides a full inner face seal for an excellent fit and helps reduce hot air buildup with the red exhalation valve. The mask protects against harmful and toxic solid and liquid particles, and it does not impair vision or speech. Latex and graphene-free, it is suitable for various industrial and hazardous environments.
Specifications
Specifications
| Aerosol Type | R - Somewhat Resistant to Oil |
| Color | White |
| Design Type | Cup/Molded |
| Disposable | Single-use |
| Filter Type | FFP3 |
| Material | Polypropylene |
| Product Type | General Purpose Particulate Respirator |
| Size | One Size |
| With Exhalation Valve | Yes |
| Latex Free | Yes |
| Show More |