Learn More
Fexofenadine-d6, MedChemExpress
Description
Fexofenadine D6 (MDL-16455 D6) is deuterium labeled is Fexofenadine, which is an antihistamine pharmaceutical agent.
Specifications
Specifications
| Chemical Name or Material | Fexofenadine-d6 |
| CAS | 548783-71-7 |
| Purity Grade Notes | Research |
| Physical Form | Powder |
| Molecular Formula | C32H33D6NO4 |
| Color | White |
| Synonym | MDL-16455-d6 Terfenadine carboxylate-d6 |
| SMILES | OC(C1=CC=CC=C1)(C2=CC=CC=C2)C3CCN(CCCC(C4=CC=C(C(C(O)=O)(C([2H])([2H])[2H])C([2H])([2H])[2H])C=C4)O)CC3 |
| Industry Type | Neuroscience-Neuromodulation |
| Molecular Weight (g/mol) | 507.69 |
| Show More |