Description
Rotating locks minimize twisting and kinking of tubing during positioning. Tapered cone seals are reliable and uncomplicated. Facilitate single-use configurations that minimize downtime
Specifications
Specifications
| Certifications/Compliance | Non-DEHP;Non-Phthalate;RoHS |
| Diameter (English) Inner | 1/8 in. |
| Material | Nylon |
| Packaging Quantity | 25/Pk. |
| Inlet | Female Luer with Panel Mount, Hose Barb |
Always test the application to verify the performance of the interconnection.