missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fc gamma RII/CD32 Rat anti-Mouse, Clone: AT130-5, Novus Biologicals™
Rat Monoclonal Antibody
451.00 EUR
Specifications
| Antigen | Fc gamma RII/CD32 |
|---|---|
| Clone | AT130-5 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Functional, Block/Neutralize |
| Applications | Western Blot, Flow Cytometry, Immunofluorescence, Functional Assay, Neutralization |
Description
Fc gamma RII/CD32 Monoclonal antibody specifically detects Fc gamma RII/CD32 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, Functional Assay, NeutralizationSpecifications
| Fc gamma RII/CD32 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, Immunofluorescence, Functional Assay, Neutralization | |
| Unconjugated | |
| Rat | |
| Immunology | |
| PBS | |
| 2213 | |
| IgG1 κ | |
| Protein A purified |
| AT130-5 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Functional, Block/Neutralize | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| CD32 antigen, CD32B, CD32Fc fragment of IgG, low affinity II, receptor for (CD32), CDw32, Fc fragment of IgG, low affinity IIb, receptor (CD32), Fc fragment of IgG, low affinity IIb, receptor for (CD32), Fc gamma RIIb, FCG2, Fc-gamma RII-b, fc-gamma-RIIb, FCGR2, fcRII-b, IGFR2, IgG Fc receptor II-b, low affinity immunoglobulin gamma Fc region receptor II-b | |
| This antibody was raised by immunising BALB/c FcgammaRIIB-/- mice with FcgammaRIIB-rat CD4 domain 3/4 fusion protein. | |
| Primary | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title