Learn More
BD™ FC Beads 5-Color Kit (CE/IVD)
Description
- Establish fluorescence compensation for a BD flow cytometer.
- BD FC beads are fluorescent beads that enable the software to calculate a fluorescence compensation matrix during setup of the flow cytometer.
- The combination of BD™ CS&T beads and BD FC beads accurately sets the voltage and compensation for each of the channels.
- In cases where two fluorophores are detected in the same channel, the BD FC beads for both colors are used during setup.
- Includes one pouch of 5 tubes for each of the following fluorochromes:
- BD Horizon™ APC-R700
- APC-H7
- BD Horizon™ V450
- BD Horizon™ V500-C
- BD Horizon Brilliant™ Violet 605
Specifications
Specifications
| Content And Storage | Store tubes at 2°C–8°C in the foil pouch. The tubes should not be frozen. Protect the tubes from exposure to light and humidity. |
| No. of Tests | 5 Tests |
| Quantity | 5 Tests |
| Stability | The beads and dilution buffer are stable until the expiration date shown on the pouch and bottle labels when stored as directed. Do not use after the expiration date. |
| Product Type | Fluorescent Polystyrene Beads |