Learn More
BD™ FC Beads 5-Color Kit (CE/IVD)
Product Code. 15951240
Change view
Click to view available options
Quantity:
5 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15951240 5 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15951240 Supplier BD™ Supplier No. 661564
Establish fluorescence compensation for a BD flow cytometer.
  • Establish fluorescence compensation for a BD flow cytometer.
  • BD FC beads are fluorescent beads that enable the software to calculate a fluorescence compensation matrix during setup of the flow cytometer.
  • The combination of BD™ CS&T beads and BD FC beads accurately sets the voltage and compensation for each of the channels.
  • In cases where two fluorophores are detected in the same channel, the BD FC beads for both colors are used during setup.
  • Includes one pouch of 5 tubes for each of the following fluorochromes:
    • BD Horizon™ APC-R700
    • APC-H7
    • BD Horizon™ V450
    • BD Horizon™ V500-C
    • BD Horizon Brilliant™ Violet 605
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store tubes at 2°C–8°C in the foil pouch. The tubes should not be frozen. Protect the tubes from exposure to light and humidity.
No. of Tests 5 Tests
Quantity 5 Tests
Stability The beads and dilution buffer are stable until the expiration date shown on the pouch and bottle labels when stored as directed. Do not use after the expiration date.
Product Type Fluorescent Polystyrene Beads