missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FBXW8 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15746441 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15746441 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15746441 Supplier Invitrogen™ Supplier No. PA558564

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: EDGFLNIWDL RTGKYPVHRF EHDARIQALA LSQDDATVAT ASAFDVVMLS PNEEGYWQIA AEFEVPKLVQ YLEIVPETRR YPVAVAAAGD LMYLLKA Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 77%.

This gene encodes a member of the F-box protein family, members of which are characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into three classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene contains a WD-40 domain, in addition to an F-box motif, so it belongs to the Fbw class. Alternatively spliced transcript variants encoding distinct isoforms have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBXW8
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FBXW8
Gene Accession No. Q8N3Y1
Gene Alias 4930438M06Rik; F-box and WD repeat domain containing 8; F-box and WD-40 domain protein 8; F-box and WD-40 domain-containing protein 8; F-box only protein 29; F-box protein FBX29; F-box/WD repeat-containing protein 8; FBW6; FBW8; Fbx29; FBXO29; FBXW6; FBXW8; LOW QUALITY PROTEIN: F-box/WD repeat-containing protein 8; MGC33534
Gene Symbols FBXW8
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FBXW8. Recombinant protein control fragment (Product #RP-102851).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26259
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.