Learn More
Scotch-Brite™ Fast Shine Microfiber Wipes 2060
Product Code. p-7213723
Click to view available options
Color:
Blue
Green
Red
Yellow
Unit Size:
Pack of 100
Product Code. 17178580
Brand: Scotch-Brite™ 7000033494
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Semi-disposable microfiber wipes with great cleaning properties. Offers quick and effective cleaning performance when used with or without chemicals on a range of surfaces.
  • Quick and effective cleaning performance when used with or without chemicals
  • Lint free cleaning on virtually any surface
  • Eliminates the need for cleaning agents in many cases
  • Ideal for cleaning furniture and sanitary ware
  • Can be used on all types of surfaces, including fragile surfaces
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cleaning Cloth
Length (Metric) 400 mm
Width (Metric) 360 mm
Material Microfiber
Color Blue
Description Semi-disposable
Dimensions (L x W) 400 x 360 mm
For Use With (Application) Quick and Effective Cleaning
Packaging Quantity 10/Pk.