Learn More
Labconco™ Fast-Freeze™ Flasks, Flask Bottom Only
Product Code. p-146987 Shop All Labconco Corporation Products
Click to view available options
Capacity (Metric):
40 mL
80 mL
120 mL
150 mL
300 mL
600 mL
900 mL
1200 mL
2000 mL
Unit Size:
Each
Product Code. 15143025
Brand: Labconco™ 7542600
Only null left
Add to basket
Add to basket
Requires
Adapters are required to attach flasks to freeze dry valve ports.
High-strength borosilicate glass bottom to withstand extreme temperatures and high vacuum.
High-strength borosilicate glass bottom to withstand extreme temperatures and high vacuum
  • Features a wide mouth opening to load fast and clean easily
  • Flat bottom reduces spills and provides better balance
  • Flasks should be filled to no more than one-third of their volume so that maximum surface area is achieved and efficient lyophilization is assured
  • Tops, bottoms, and filter paper are available separately as replacement components
Specifications
Diameter (English) Outer 2.32 in.
Capacity (Metric) 300 mL
Material Borosilicate Glass
Height (English) 5.7 in.
Height (Metric) 14.5 cm
Product Type Flask
Weight (English) 1.5 lb.
Disposable Reusable
Mouth Style Wide
Bottom Shape Flat
Diameter (Metric) Outer 5.9 cm
Weight (Metric) 0.7 kg