missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM78B Polyclonal Antibody

Description
Recommended positive controls: Jurkat. Predicted reactivity: Mouse (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
FAM78B (Family With Sequence Similarity 78 Member B) is a Protein Coding gene. An important paralog of this gene isFAM78A.
Specifications
Specifications
| Antigen | FAM78B |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | FAM78B |
| Gene Accession No. | Q5VT40 |
| Gene Alias | C030014K22Rik; C030020L09Rik; Fam78b; family with sequence similarity 78 member B; family with sequence similarity 78, member B; protein FAM78B |
| Gene Symbols | FAM78B |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?