missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FAM65A Polyclonal Antibody
GREENER_CHOICE

Product Code. 16588641
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16588641 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16588641 Supplier Invitrogen™ Supplier No. PA595803

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: LSLRQPGLAE AVCVKFLEDA LGQKLPRRPQ PGPGEQLTVF QFWSFVETLD SPTMEAYVTE TAEEVLLVRN LNSDDQAVVL KALRLAPEGR LRRDGLRALS SLLVHGNNKV MAAVSTQLRS LSLGPTFRER ALLCFLDQLE DEDVQTRVAG CLALGCIKAP EGIEPLVYLC QTDTEAVREA ARQSLQQCGE EGQSAHRRLE ESLDALPRIF GPGSMASTAF; Positive Samples: Mouse kidney; Cellular Location: Cytoplasm.

Downstream effector protein for Rho-type small GTPases that plays a role in cell polarity and directional migration (PubMed:27807006). Acts as an adapter protein, linking active Rho proteins to STK24 and STK26 kinases, and hence positively regulates Golgi reorientation in polarized cell migration upon Rho activation (PubMed:27807006). Involved in the subcellular relocation of STK26 from the Golgi to cytoplasm punctae in a Rho- and PDCD10-dependent manner upon serum stimulation (PubMed:27807006). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM65A
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene RIPOR1
Gene Accession No. Q68FE6
Gene Alias 2310066E14Rik; AI837433; Fam65a; family with sequence similarity 65 member A; family with sequence similarity 65, member A; FLJ13725; Kiaa1930; protein FAM65A; Rho family-interacting cell polarization regulator 1; RIPOR1
Gene Symbols RIPOR1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1004-1223 of human FAM65A (NP_0011804511).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 75687
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.