missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM172A Polyclonal Antibody

Description
Peptide sequence: SSDSSDEPAE KRERKDKVSK ETKKRRDFYE KYRNPQREKE MMQLYIRENG Sequence homology: Dog: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%; Yeast: 83%.
FAM172A is a protein coding gene.
Specifications
Specifications
| Antigen | FAM172A |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | FAM172A |
| Gene Accession No. | Q8WUF8 |
| Gene Alias | 1110033M05Rik; 2610318O14Rik; 53-E6; 9430037D06Rik; AF064782; C5orf21; Cotranscriptional regulator FAM172A; Fam172a; family with sequence similarity 172 member A; family with sequence similarity 172, member A; LOW QUALITY PROTEIN: protein FAM172A; pEN87; Protein FAM172A; RGD1305526 |
| Gene Symbols | FAM172A |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?