Learn More
Memmert™ Factory Calibration for UN160plus
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
UN160plus Oven
Unit Size:
Each
Description
- For a freely selectable temperature value
Specifications
Specifications
| For Use With (Equipment) | UN160plus Oven |
| Product Type | Factory Calibration |