Learn More
Memmert™ Factory Calibration (3 Temperatures) for UN450
Product Code. 17570176 Shop All Memmert Products
Change view
Click to view available options
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. unitSize
17570176 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17570176 Supplier Memmert™ Supplier No. UN450D00128
Calibration at Memmert factory for three temperature value.
  • For three temperature values: 100°C, 160°C, and 220°C
Specifications
For Use With (Equipment) UN450 Oven
Description Measuring point chamber center
Calibration Type Certificate for 3 temperatures
Product Type Standard Works Calibration Certificate