missing translation for 'onlineSavingsMsg'
Learn More

Fabomotizole hydrochloride, MedChemExpress

Product Code. 16446248
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16446248

Brand: Medchem Express HY14895A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Fabomotizole hydrochloride (CM346 hydrochloride) is an anxiolytic drug. Fabomotizole hydrochloride produces anxiolytic and neuroprotective effects without any muscle relaxant actions.

Fabomotizole hydrochloride (CM346 hydrochloride) is an anxiolytic drug. Fabomotizole hydrochloride produces anxiolytic and neuroprotective effects without any muscle relaxant actions.

Specifications

Chemical Name or Material Fabomotizole hydrochloride
CAS 173352-39-1
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C15H22ClN3O2S
Quantity 10 mg
Synonym CM346 hydrochloride
Solubility Information H2O : 100 mg/mL (290.81 mM; Need ultrasonic) ∣DMSO : ≥ 44 mg/mL (127.96 mM)
SMILES CCOC1=CC=C2C(N=C(SCCN3CCOCC3)N2)=C1.Cl
Molecular Weight (g/mol) 343.87
Formula Weight 343.87
Percent Purity 99.77%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.