missing translation for 'onlineSavingsMsg'
Learn More
Learn More
F4/80, Rat anti-Mouse, Clone: Cl:A3-1 (recombinant version), Novus Biologicals™
Rat Monoclonal Antibody
261.00 EUR - 595.00 EUR
Specifications
| Antigen | F4/80 |
|---|---|
| Clone | Cl:A3-1 (recombinant version) |
| Concentration | 1 mg/mL |
| Dilution | Flow Cytometry, Immunohistochemistry |
| Applications | Flow Cytometry, Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18308000
|
Novus Biologicals
NBP2-81030-0.025MG |
0.025 mg |
261.00 EUR
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18390810
|
Novus Biologicals
NBP2-81030-0.2MG |
0.2 mg |
595.00 EUR
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
F4/80 Monoclonal specifically detects F4/80 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry.Specifications
| F4/80 | |
| 1 mg/mL | |
| Flow Cytometry, Immunohistochemistry | |
| Unconjugated | |
| Rat | |
| Adaptive Immunity, GPCR, Immune System Diseases, Immunology, Innate Immunity, Myeloid derived Suppressor Cell | |
| PBS | |
| 13733 | |
| Thioglycollate stimulated peritoneal macrophages of mouse origin. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Cl:A3-1 (recombinant version) | |
| Flow Cytometry, Immunohistochemistry | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| ADGRE1, cell surface glycoprotein F4/80, DD7A5-7, EGF-like module containing, mucin-like, hormone receptor-like sequence 1, EGF-like module receptor 1, EGF-like module-containing mucin-like hormone receptor-like 1, EGF-TM7, EMR1 hormone receptor, F4/80, Gpf480, Ly71, lymphocyte antigen 71, Mouse EMR1, Rat EMR1, TM7LN3 | |
| Adgre1 | |
| IgG2b κ | |
| Protein A purified | |
| This antibody recognises the mouse F4/80 antigen, a 160kD glycoprotein, expressed by murine macrophages. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title