missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EZH2 Mouse anti-Human, Mouse, Rat, Clone: 1F10A12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66476-1-IG-20UL
This item is not returnable.
View return policy
Description
This antibody detected EZH2 protein as a single band with a molecular weight (MW) of 91-100 kDa in multiple cell lines. The phosphorylation may result in the higher molecular weight (calculated MW as 80-86 kDa).
FANCD2 (Fanconi anemia group D2 protein) is required for maintece of chromosomal stability. It promotes accurate and efficient pairing of homologs during meiosis. FANCD2 is involved in the repair of DNA double-stranded breaks, both by homologous recombination and single-stranded annealing. It is required for the targeting/stabilization of BLM to non-centromeric abnormal structures induced by replicative stress. FANCD2 promotes BRCA2/FANCD1 loading onto damaged chromatin. Defects in the FANCD2 gene result in Fanconi anemia complementation group D2. This disorder affects all bone marrow elements resulting in anemia, leukopenia, and thrombopenia.Specifications
| EZH2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q15910, Q61188 | |
| Ezh2 | |
| EZH2 Fusion Protein ag16789 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 1F10A12 | |
| Unconjugated | |
| EZH2 | |
| Enhancer of zeste homolog 2, ENX 1, EZH1, EZH2, KMT6, Lysine N methyltransferase 6 | |
| Mouse | |
| Protein G | |
| RUO | |
| 14056, 2146, 312299 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction