Learn More
Invitrogen™ EYA1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540545
This item is not returnable.
View return policy
Description
Peptide sequence: QDYPSYPSFG QGQYAQYYNS SPYPAHYMTS SNTSPTTPST NATYQLQEPP Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.
This gene encodes a member of the eyes absent family of proteins. The encoded protein may play a role in the developing kidney, branchial arches, eye, and ear. Mutations of this gene have been associated with branchiootorenal dysplasia syndrome, branchiootic syndrome, and sporadic cases of congenital cataracts and ocular anterior segment anomalies. A similar protein in mice can act as a transcriptional activator. Four transcript variants encoding three distinct isoforms have been identified for this gene.
Specifications
| EYA1 | |
| Polyclonal | |
| Unconjugated | |
| EYA1 | |
| BOP; bor; BOS1; dog eared; EYA transcriptional coactivator and phosphatase 1; eya1; eya1 protein; Eyes absent; eyes absent 1; eyes absent 1 homolog; eyes absent 1 homolog (Drosophila); eyes absent homolog 1; MGC141875; OFC1; OTTHUMP00000195053; wu:fc13c10; zgc:100770 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 2138 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q99502 | |
| EYA1 | |
| Synthetic peptide directed towards the middle region of human EYA1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |