Learn More
Lenz Laborglasintrumente Extractors for Solids
Click to view available options
Capacity (Metric):
70 mL
100 mL
250 mL
Unit Size:
Each
Description
- Extractor head with glass thimbles for solids
- Made of DURAN™ tubing
- Dimroth condenser (details on page 71)
- With thread GL 14
- Round bottom flask
Specifications
Specifications
| Capacity (Metric) | 70 mL |
| Material | Borosilicate Glass 3.3 |
| Standard Taper Joint Side | NS 45/40 |