Learn More
Lenz Laborglasintrumente Extractor Heads for Specific Light Solvents
Product Code. p-7222481
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
Unit Size:
Each
Product Code. 10385971
Brand: Lenz Laborglasintrumente 05432137
Only null left
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • For the extraction of liquids with specific light solvents
  • With insert filter (porosity: 0)
  • Made of DURAN™ tubing
Specifications
Capacity (Metric) 100 mL
Material Fritte P0
Standard Taper Joint NS 45/40