Learn More
Memmert™ External Measuring Instrument
Product Code. 17100085 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
HPP110eco
HPP260eco
HPP410eco
HPP750eco
IPP110eco
IPP260eco
IPP410eco
IPP750eco
Unit Size:
Each
Product Code. 17100085
Brand: Memmert™ IPP110ecoB04713
Only null left
Add to basket
Add to basket
External measuring instrument with sensors for daylight and UV-light
  • Sensors for daylight and UV-light
Specifications
For Use With (Equipment) IPP110eco
Includes Sensors for daylight and UV-light