Learn More
Memmert™ Extension of DAkkS Factory Calibration for SN110plus
Product Code. 17568475 Shop All Memmert Products
Click to view available options
Unit Size:
Each
Product Code. 17568475
Brand: Memmert™ SN110plusE39697
Extension of DAkkS factory calibration by an additional freely selectable temperature for Memmert Sterilizers.
  • Extension of DAkkS calibration at Memmert factory by an additional freely selectable temperature value according to method C (DKD-R 5-7)
Specifications
For Use With (Equipment) SN110plus Sterilizer
Product Type Factory Calibration Extension