Learn More
Benchmark Scientific exchangeable Microplate Block
Product Code. 16450251
Change view
Click to view available options
Holds:
1 Standard Microplate
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds unitSize
16450251 1 Standard Microplate Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16450251 Supplier Benchmark Scientific Supplier No. H5000MP
Only null left
Add to basket
Add to basket
For MultiTherm™ Temperature Controlled Vortexer, holds 1 standard microplate
  • For Benchmark MultiTherm™ Temperature Controlled Vortexer
  • Holds 1 standard microplate
Specifications
Holds 1 Standard Microplate
Product Type Exchangable Block
For Use With (Equipment) Benchmark MultiTherm™
No. of Positions 1