Learn More
Benchmark Scientific exchangeable Block
Description
- For Benchmark MultiTherm™ Temperature Controlled Vortexer
- Holds 12 HPLC/Autosampler Vials (12 x 32 mm)
Specifications
Specifications
| Holds | HPLC/Autosampler Vials |
| Tube Size | 32 mm |
| Product Type | Exchangable Block |
| For Use With (Equipment) | HPLC/Autosampler Vials |
| No. of Positions | 12 |