Learn More
Epredia™ Excelsior™ AS Tissue Processor Accessories
Product Code. 12688156 Shop All Epredia Products
Click to view available options
Product Type:
Air System Duct Adapter Kit
Basket Carrying Container
Downdraft Extrctn Duct Adapter
Monitor Clamp Kit
Organized Basket, 222 cassette
Organized Basket, 300 cassette
Potassium Permanganate Filter
Storage Bottle
Vapor Filter (Charcoal)
Waste Wax Drawer Kit
Unit Size:
Each
Pack of 6
Product Code. 12688156
Brand: Epredia™ A78410029
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Maintain the Epredia™ Excelsior™ AS Tissue Processor with Epredia™ Excelsior AS Tissue Processor Accessories.
For use with Shandon Excelsior ES Tissue Processor
TRUSTED_SUSTAINABILITY
Specifications
Product Type Monitor Clamp Kit
For Use With (Equipment) Excelsior ES Tissue Processor