Learn More
Epredia™ Excelsior™ AS Tissue Processor Accessories
Shop All Epredia Products
Click to view available options
Product Type:
Air System Duct Adapter Kit
Basket Carrying Container
Downdraft Extrctn Duct Adapter
Monitor Clamp Kit
Organized Basket, 222 cassette
Organized Basket, 300 cassette
Potassium Permanganate Filter
Storage Bottle
Vapor Filter (Charcoal)
Waste Wax Drawer Kit
Unit Size:
Each
Pack of 6
Description
For use with Shandon Excelsior ES Tissue Processor
Specifications
Specifications
| Product Type | Basket Carrying Container |
| For Use With (Equipment) | Excelsior AS Tissue Processor |
| Material | Plastic |