Learn More
Abnova™ EWSR1/ATF1 DY Translocation FISH Probe
Product Code. 16043575
Click to view available options
Quantity:
100 μL
200 μL
Unit Size:
100µL
200µL
Product Code. 16043575
Brand: Abnova™ FT0022.100uL
Labeled FISH probes for identification of gene translocation using Fluoresecent In Situ Hybridization Technique.
Specifications
Content And Storage Store at 4°C in the dark
Format Tube
For Use With (Application) Fluorescent In Situ Hybridization (Cell)
Gene Ewing sarcoma breakpoint region 1
Gene Alias EWS
Gene Symbol EWSR1
Gene ID (Entrez) 2130;466
Product Type DNA/RNA
Quantity 100 μL
Form Liquid