Learn More
Thermo Scientific™ Evolution™ UV-Visible Spectrophotometers Smart Thermostatted 7-Cell Rotary Changer
Product Code. p-6268000 Shop All Thermo Scientific Products
Click to view available options
For Use With (Equipment):
Evolution Spectrophotometers
Type:
7-Cell Rotary Changer
Unit Size:
Each
Product Code. 12306583
Brand: Thermo Scientific™ 840214900
Efficiently control temperature with a circulating water bath using the Thermo Scientific™ Evolution™ UV-Visible Spectrophotometers Smart Thermostatted 7-Cell Rotary Changer.
Efficiently control temperature with a circulating water bath using the Thermo Scientific™ Evolution™ UV-Visible Spectrophotometers Smart Thermostatted 7-Cell Rotary Changer. Combined with a liquid recirculator, this cell changer provides control of sample temperature for up to 7 samples or standards.
  • Easy-to-use sample positioning accessory for use with Thermo Scientific™ Evolution™ 200 series and 300 spectrophotometers.
  • Accessory control fully integrated in Thermo Scientific™ INSIGHT™ and Thermo Scientific™ VISION™ software.
  • Automatic software recognition, smart alignment and serial number reporting
  • Includes carousel and motor base.
  • When placed in both sample and reference beams (Evolution 300 only), this cell changer allows comparison of up to 7 samples with 7 unique references.
  • Monitor the temperature in a cell with Temperature probe accessory for Smart accessories, catalog number 0011401.
  • When deploying this accessory in a regulated or QC environment, a full-system qualification package is available to assure compliance of instrument and accessory as a complete unit.
TRUSTED_SUSTAINABILITY
Specifications
Compatibility Compatible with temperature probe accessory for Smart accessories, catalog#10011401
Configuration Details Evolution One/One Plus- sample position only, Evolution Pro- sample and reference positions
Type 7-Cell Rotary Changer
For Use With (Equipment) Evolution Spectrophotometers
Includes Carousel and motor base
Thermostatting Thermostatting with flowing liquid from an external temperature controlled recirculator