missing translation for 'onlineSavingsMsg'
Learn More

EVI1 Recombinant Superclonal™ Antibody

Product Code. 15815783
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15815783 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15815783 Supplier Invitrogen Supplier No. 711037

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

EVI1 Recombinant Polyclonal Antibody

EVI1 Recombinant Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen EVI1
Applications Western Blot, Immunocytochemistry
Classification Recombinant Superclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene MECOM
Gene Accession No. Q03112
Gene Alias AML1-EVI-1; AML1-EVI-1 fusion protein; D630039M04Rik; ecotropic viral integration site 1; ecotropic virus integration site 1 protein; Ecotropic virus integration site 1 protein homolog; EVI1; Evi-1; Histone-lysine N-methyltransferase MECOM; Jbo; Mds; MDS1; MDS1 and EVI1 complex locus; MDS1 and EVI1 complex locus protein; MDS1 and EVI1 complex locus protein EVI1; MDS1 and EVI1 complex locus protein MDS1; Mds1-Evi1; Mecom; MGC163392; MGC97004; myelodysplasia syndrome 1 homolog; Myelodysplasia syndrome 1 protein; Myelodysplasia syndrome 1 protein homolog; myelodysplasia syndrome-associated protein 1; oncogene EVI1; OTTHUMP00000213163; OTTHUMP00000213164; OTTHUMP00000213165; OTTHUMP00000213187; OTTHUMP00000213192; PR domain 3; PRDM3; RUSAT2; zinc finger protein Evi1; Znfpr1b1
Gene Symbols MECOM
Host Species Rabbit
Immunogen Peptide corresponding to Human MECOM (aa 367-384).
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2122
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.