Learn More
Fisherbrand™ Sealing Mat for 96-Well Deep Square Nonsterile Microplates
Product Code. 11521963
Encompass
Change view
Click to view available options
Product Type:
96 Well Square Sealing Cap
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type unitSize
11521963 96 Well Square Sealing Cap Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11521963 Supplier Fisherbrand™ Supplier No. 219004
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Fisherbrand™ Sealing Mat for 96-Well Deep Square Nonsterile Microplates is a reusable EVA sealing cap that fits onto the top of the plate to retain the sample in the well during transit or to stop the well and its contents from becoming contaminated.
  • For friction sealing of plates during transport or storage
  • Cap mats are an economic alternative to thermal or adhesive seals
  • Cap mats can be sterilized and reused
  • Constructed of high quality ethylene-vinyl acetate (EVA)
  • Rnase/Dnase free
Specifications
Color White
Material EVA
Sterility Non-sterile
No. of Wells 96
No. per Pack 50
Product Type 96 Well Square Sealing Cap
Well Shape Square
Plate Compatibility 96-well Square Deepwell Plate