Learn More
Thermo Scientific™ Eutech™ Cyberscan Electrode Holder for ATC probes and pH electrodes
Product Code. p-7114746
Click to view available options
For Use With (Equipment):
16mm ATC Probes and pH Electrodes
Unit Size:
Each
Product Code. 11509264
Brand: Thermo Scientific™ 15X000702
Optimize your Eutech™ meters and electrodes with these accessories and replacement parts.
Optimize your Eutech™ meters and electrodes with these accessories and replacement parts. The Eutech™ Cyberscan Electrode Holder is used for Eutech™ ATC probes and pH electrodes in between sample readings.
Features:
  • Holds 12mm diameter or 16mm diameter electrodes
TRUSTED_SUSTAINABILITY
Specifications
Type Electrode Holder
For Use With (Equipment) 16mm ATC Probes and pH Electrodes