Learn More
Coverguard™ EUROWELD Gloves
Description
- CE marked
- EPI PPE Category II
- Cut and sewn cowhide leather
- Safety cuff with split - can be worn long or folded down
- Wing thumb provides flexibility
- Vein patch protector
- Protection against heat up to 250°C for 15 seconds
- Not for use with chemicals
Specifications
Specifications
| Certifications/Compliance (Europe) | EPI PPE Cat. II, EN 420:2003 + A1:2009, EN 388:2016, EN 407:2004 |
| Applications | Industrial |
| Color | Gray |
| Product Type | Protective Gloves |
| Size | 10 |
| Thickness (English) | 39 to 51 mil |
| Thickness (Metric) | 1.0 to 1.3 mm |
| EN388 Standard | EN 388:2016 |
| Glove Cuff Style | Open |
| Model | EUROWELD 2507 |
| Show More |