Learn More
IKA™ EUROSTAR 20 High Speed Digital Overhead Stirrer
GREENER_CHOICE
Product Code. 13430577 Shop All IKA Products
Change view
Click to view available options
Plug Type:
EU
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Plug Type unitSize
13430577 EU Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13430577 Supplier IKA™ Supplier No. 0004028600
Only null left
Add to basket
Add to basket
High speed laboratory stirrer designed for intensive stirring tasks for quantities up to 20 L (H2O).
  • Safety circuits installed ensures automatic cut-off in an anti-stall or overload conditions
  • Continuous comparison of shaft speed to desired speed is maintained and variations are adjusted automatically
  • Guarantees a constant speed even with changes in viscosities of the sample
  • Precision shaft is 280 mm long and has a maximum immersion depth of 220 mm
  • Digital speed display
  • Infinitely adjustable speed
  • Cone seat for precision shaft
  • Overload protection
  • Short-term overload operation
  • Slim casing
  • Quiet operation
  • Error code display
TRUSTED_SUSTAINABILITY
Specifications
Speed 150 to 6000 rpm
Capacity 20 L
Display 7 Segment LED
Voltage 230 V
Wattage 176 w
Height (Metric) 325 mm
Width (Metric) 86 mm
Electrical Requirements 230V, 50/60Hz, 176w
Frequency 50/60 Hz
Plug Type EU
Material Alu-Cast Coating/Thermoplastic Polymer
Model EUROSTAR 20 High Speed Digital
Temperature Range (Metric) 5°C to 40°C
Torque 20 Ncm
Viscosity 10,000 mPas