missing translation for 'onlineSavingsMsg'
Learn More

Etonogestrel Sulfate Sodium Salt, TRC

Product Code. 30378125
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30378125 50 mg 50mg
30383089 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30378125 Supplier Toronto Research Chemicals Supplier No. TRCE93363550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Etonogestrel Sulfate Sodium Salt
Analyte or Component Names Etonogestrel Sulfate Sodium Salt
Shipping Condition Room Temperature
Molecular Formula C22 H27 Na O5 S
InChI Formula InChI=1S/C22H28O5S.Na/c1-4-21-13-14(3)20-17-9-7-16(23)12-15(17)6-8-18(20)19(21)10-11-22(21,5-2)27-28(24,25)26;/h2,12,17-20H,3-4,6-11,13H2,1H3,(H,24,25,26);/q;+1/p-1/t17-,18-,19-,20+,21-,22-;/m0./s1
Solution Type Neat
SMILES CC[C@]12CC(=C)[C@H]3[C@@H](CCC4=CC(=O)CC[C@H]34)[C@@H]1CC[C@@]2(OS(=O)(=O)O[Na])C#C
IUPAC Name [(8S,9S,10R,13S,14S,17R)-13-ethyl-17-ethynyl-11-methylidene-3-oxo-2,6,7,8,9,10,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl]oxysulfonyloxysodium
Industry Type Pharmaceutical
Molecular Weight (g/mol) 426.502
Quantity 50 mg
Formula Weight 426.148
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.