Learn More
Etomoxir sodium salt, MedChemExpress
Product Code. 16410538
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16410538 5 mg 5mg
16468068 10 mg 10mg
16453987 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16410538 Supplier Medchem Express Supplier No. HY50202A
MedChemExpress Etomoxir((R)-(+)-Etomoxir) sodium salt is an irreversible inhibitor of carnitine palmitoyltransferase 1a (CPT-1a), inhibits fatty acid oxidation (FAO) through CPT-1a and inhibits palmitate β-oxidation in human, rat and guinea pig.
Etomoxir((R)-(+)-Etomoxir) sodium salt is an irreversible inhibitor of carnitine palmitoyltransferase 1a (CPT-1a), inhibits fatty acid oxidation (FAO) through CPT-1a and inhibits palmitate β-oxidation in human, rat and guinea pig.
Specifications
Chemical Name or Material Etomoxir sodium salt
CAS 828934-41-4
Color White
Purity Grade Notes Research
Molecular Formula C15H18ClNaO4
Quantity 5 mg
Synonym (R)-(+)-Etomoxir sodium salt
Solubility Information DMSO : 100 mg/mL (311.78 mM; Need ultrasonic) ∣H2O : 5 mg/mL (15.59 mM; Need ultrasonic)
SMILES ClC1=CC=C(OCCCCCC[C@@]2(CO2)C(O[Na])=O)C=C1
Molecular Weight (g/mol) 320.74
Formula Weight 320.74
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death