Learn More
EDTA disodium salt solution with zinc complex added (solution B), 1 ml solution = 1 German degree of hardness in 100 ml of water, for complexometry, Honeywell Fluka™
Shop All Honeywell Fluka ProductsDescription
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
Specifications
| Chemical Name or Material | Ethylenediaminetetraacetic acid disodium salt solution with zinc complex added (solution B) |
| CAS | 7646-85-7, 60-00-4, 7732-18-5 |
| CAS Max % | 0.7, 0.02 |
| Molecular Formula | Cl2Zn |
| UN Number | NONH for all modes of transport |
| InChI Key | JIAARYAFYJHUJI-UHFFFAOYSA-L |
| SMILES | [Cl-].[Cl-].[Zn++] |
| IUPAC Name | zinc(2+) dichloride |
| Molecular Weight (g/mol) | 136.28 |
| Packaging | Poly Bottle |
| Show More |