missing translation for 'onlineSavingsMsg'
Learn More

Ethyl trifluoropyruvate, 97%

Product Code. p-7025231
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11419037

Brand: Thermo Scientific Alfa Aesar B20611.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13081-18-0 | C5H5F3O3 | 170.09 g/mol

Ethyl trifluoropyruvate acts as a building block in synthetic chemistry for the synthesis of fluorine containing compounds. It is also used as a reagent for the preparation of potential biologically active compounds like highly enantioselective organocatalytic hydroxyalkylation of indoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl trifluoropyruvate acts as a building block in synthetic chemistry for the synthesis of fluorine containing compounds. It is also used as a reagent for the preparation of potential biologically active compounds like highly enantioselective organocatalytic hydroxyalkylation of indoles.

Solubility
Miscible with dichloromethane.

Notes
Incompatible with strong oxidizing agents, reducing agents, acids and alkalis (bases). Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13081-18-0
Molecular Formula C5H5F3O3
Molecular Weight (g/mol) 170.09
MDL Number MFCD00114935
InChI Key KJHQVUNUOIEYSV-UHFFFAOYSA-N
Synonym ethyl trifluoropyruvate, ethyltrifluoropyruvate, ethyl 3,3,3-trifluoropyruvate, trifluoropyruvic acid ethyl ester, propanoic acid, 3,3,3-trifluoro-2-oxo-, ethyl ester, ethyl 2-oxo-3,3,3-trifluoropropanoate, 3,3,3-trifluoro-2-oxopropanoic acid ethyl ester, ethyl trifluorpyruvate, ethyl-trifluoropyruvate, pubchem2013
PubChem CID 2737239
IUPAC Name ethyl 3,3,3-trifluoro-2-oxopropanoate
SMILES CCOC(=O)C(=O)C(F)(F)F

Specifications

Density 1.4
Boiling Point 95°C to 98°C
Flash Point 27°C (80°F)
Refractive Index 1.3405
Quantity 5 g
UN Number UN3272
Beilstein 2087388
Solubility Information Miscible with dichloromethane.
Formula Weight 170.09
Percent Purity 97%
Chemical Name or Material Ethyl trifluoropyruvate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.