missing translation for 'onlineSavingsMsg'
Learn More

Ethyl piperidine-1-acetate, 98%, Thermo Scientific™

Product Code. 11480254
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11480254 100 g 100g
11460254 5 g 5g
11470254 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11480254 Supplier Thermo Scientific Alfa Aesar Supplier No. A13663.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23853-10-3
Molecular Formula C9H17NO2
Molecular Weight (g/mol) 171.24
MDL Number MFCD00134496
InChI Key LXZHLNWWDDNIOJ-UHFFFAOYSA-N
PubChem CID 643466
IUPAC Name ethyl 2-piperidin-1-ylacetate
SMILES CCOC(=O)CN1CCCCC1

Specifications

Density 0.984
Boiling Point 109°C to 111°C (25 mmHg)
Flash Point 91°C (195°F)
Refractive Index 1.454
Quantity 100 g
Beilstein 123142
Synonym ethyl 1-piperidineacetate, ethyl piperidin-1-ylacetate, ethyl 2-piperidin-1-yl acetate, ethyl piperidine-1-acetate, 1-piperidineacetic acid, ethyl ester, ethyl 2-piperidylacetate, ethyl1-piperidineacetate, acmc-20al1z, ethyl 1-piperidine acetate, ethyl 2-piperidinylacetate
Formula Weight 171.24
Percent Purity 98%
Chemical Name or Material Ethyl piperidine-1-acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.