Learn More
Ethyl hexanoate, 99%
Product Code. 11451748
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11451748 2500 g 2500g
11431748 100 g 100g
11441748 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11451748 Supplier Thermo Scientific Alfa Aesar Supplier No. A13985.0E
Request Bulk or Custom Format
CAS: 123-66-0 | C8H16O2 | 144.214 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 123-66-0
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.214
MDL Number MFCD00009511
InChI Key SHZIWNPUGXLXDT-UHFFFAOYSA-N
Synonym ethyl caproate, ethyl hexoate, hexanoic acid, ethyl ester, hexanoic acid ethyl ester, capronic ether absolute, caproic acid ethyl ester, ethyl n-hexanoate, ethyl butyl acetate, caproate, acetic acid, butyl-, ethyl ester
PubChem CID 31265
ChEBI CHEBI:86055
IUPAC Name ethyl hexanoate
SMILES CCCCCC(=O)OCC
Specifications
Melting Point -67°C
Density 0.873
Boiling Point 166°C
Flash Point 49°C (121°F)
Refractive Index 1.4075
Quantity 2500 g
UN Number UN3272
Merck Index 14,3777
Formula Weight 144.21
Percent Purity 99%
Chemical Name or Material Ethyl hexanoate