Learn More
Ethyl-Beta-D-glucuronide, Lipomed
Product Code. 30458081
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30458081 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30458081 Supplier Lipomed Supplier No. LPMEGL332100
Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.
Ethyl-Beta-D-glucuronide
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Ethyl-beta-D-glucuronide
CAS 17685-04-0
Analyte or Component Names Ethyl-beta-D-glucuronide
Shipping Condition Room Temperature
Molecular Formula C8 H14 O7
Synonym β-D-Glucopyranosiduronic acid, ethyl (9 CI, ACI), Ethyl β-D-glucopyranosiduronic acid (ACI), Glucopyranosiduronic acid, ethyl, β-D- (6 CI, 8 CI), BA 2829, Ethyl glucuronide, Ethyl β-D-glucosiduronic acid, Ethyl β-D-glucuronide
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES CCO[C@@H]1 O[C@@H]([C@@H](O)[C@H](O)[C@H]1 O)C(=O)O
IUPAC Name (2 S,3 S,4 S,5 R,6 R)-6-ethoxy-3,4,5-trihydroxyoxane-2-carboxylic acid
Industry Type Pharmaceutical
Molecular Weight (g/mol) 222.193
Quantity 100 mg
Formula Weight 222.074 g/mol